EMAIL THIS PAGE TO A FRIEND
47930-U Supelco

QTM PAH Mix

2000 μg/mL each component in methylene chloride, analytical standard

Purchase

Properties

grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 mL
concentration   2000 μg/mL each component in methylene chloride
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
format   multi-component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

Product #

Add to Cart

Acenaphthene    
Acenaphthylene    
Anthracene    
Benz[a]anthracene    
Benzo[b]fluoranthene    
Benzo[ghi]perylene    
Benzo[a]pyrene    
2-Bromonaphthalene    
Chrysene    
Dibenz[a,h]anthracene    
Fluoranthene    
Fluorene    
Indeno[1,2,3-cd]pyrene    
Naphthalene    
Phenanthrene    
Pyrene    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

CRM47930 QTM PAH Mix, certified reference material, 2000 μg/mL each component in dichloromethane, ampule of 1 mL

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?