EMAIL THIS PAGE TO A FRIEND
47959 Supelco

Azobenzene solution

2000 μg/mL in methanol, analytical standard

  • CAS Number 103-33-3

  • Empirical Formula (Hill Notation) C12H10N2

  • Molecular Weight 182.22

Purchase

Properties

Related Categories A-B, A-C, Additional Standards, Alpha Sort, Analytical Standards,
grade   analytical standard
packaging   ampule of 1 mL
concentration   2000 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Cleaning Products
Cosmetics
Environmental
Food and Beverages
Personal Care
format   single component solution
storage temp.   2-8°C
InChI   1S/C12H10N2/c1-3-7-11(8-4-1)13-14-12-9-5-2-6-10-12/h1-10H
InChI key   DMLAVOWQYNRWNQ-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

GC Analysis of a 32-Component Anilines Mix on SLB®-IL59

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 32-Component Anilines Mix on SLB®-IL59
Keywords: Chromatography, Gas chromatography, Mass selective detector

US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.
Keywords: Chromatography, Gas chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?