• USA Home
  • 47962 - Aroclor 1016 solution

EMAIL THIS PAGE TO A FRIEND
47962 Supelco

Aroclor 1016 solution

certified reference material, 500 mg/kg in transformer oil

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Aroclors, Aroclors, PCBs, and Dioxins,
grade   certified reference material
  TraceCERT®
packaging   ampule of 5 mL
concentration   500 mg/kg in transformer oil
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
format   single component solution
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2315 9 / PGII
WGK Germany 
3
Flash Point(F) 
294.8 °F
Flash Point(C) 
146 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?