• USA Home
  • 47977 - EPA OLM04 Pesticide Standard Mix A-1

EMAIL THIS PAGE TO A FRIEND
47977 Supelco

EPA OLM04 Pesticide Standard Mix A-1

in hexane: toluene (99:1) (varied), analytical standard

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, CLP Standards, Chromatography,
grade   analytical standard
feature   standard type calibration
packaging   ampule of 1 mL
concentration   in hexane: toluene (99:1) (varied)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
format   multi-component solution
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

Product #

Add to Cart

α-BHC    
1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane 1 μg/mL    
4,4′-DDT 1 μg/mL    
Decachlorobiphenyl 1 μg/mL    
Dieldrin 1 μg/mL    
α-Endosulfan .5 μg/mL    
Endrin 1 μg/mL    
Heptachlor .5 μg/mL    
Lindane .5 μg/mL    
Methoxychlor 5 μg/mL    
2,4,5,6-Tetrachloro-m-xylene .5 μg/mL    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1208 - class 3 - PG 2 - Hexanes
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?