EMAIL THIS PAGE TO A FRIEND
48035 Supelco

o-Toluidine solution

2000 μg/mL in methanol, analytical standard

Synonym: 2-Methylaniline

Purchase

Properties

Related Categories 2002/61/EC, Additional Standards, Alpha Sort, Amines, Analytical Standards,
Quality Level   100
grade   analytical standard
packaging   ampule of 1 mL
concentration   2000 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
format   single component solution
storage temp.   2-8°C
SMILES string   Cc1ccccc1N
InChI   1S/C7H9N/c1-6-4-2-3-5-7(6)8/h2-5H,8H2,1H3
InChI key   RNVCVTLRINQCPJ-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

GC Analysis of Anilines on Equity®-5

From our library of Articles, Sigma-Aldrich presents GC Analysis of Anilines on Equity®-5
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Semivolatiles on SLB®-5ms after SPME using 60 μm Carbowax® Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Semivolatiles on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
Keywords: Chromatography, Gas chromatography, Purification, Solid phase microextractions

GC Analysis of a 16-Component Anilines Mix on SLB®-IL59

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 16-Component Anilines Mix on SLB®-IL59
Keywords: Chromatography, Gas chromatography

GC Analysis of a 32-Component Anilines Mix on SLB®-IL59

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 32-Component Anilines Mix on SLB®-IL59
Keywords: Chromatography, Gas chromatography, Mass selective detector

HPLC Analysis of Toluidine Isomers on Ascentis® Si (Hexane: 2-Propanol)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Toluidine Isomers on Ascentis® Si (Hexane: 2-Propanol)
Keywords: Chromatography, High performance liquid chromatography

HPLC Analysis of Toluidine Isomers on Ascentis® Si (Silica), (80:20 Hexane:Ethyl Acetate)

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Toluidine Isomers on Ascentis® Si (Silica), (80:20 Hexane:Ethyl Acetate)
Keywords: Chromatography, High performance liquid chromatography

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?