EMAIL THIS PAGE TO A FRIEND
48058 Supelco

EPA HC Purgeable B

certified reference material, 2000 μg/mL each component in methanol

Purchase

Properties

Quality Level   100
grade   certified reference material
Agency/Method   EPA 624
feature   standard type calibration
packaging   ampule of 1 mL
concentration   2000 μg/mL each component in methanol
Featured Industry   Environmental
format   multi-component solution
storage temp.   2-8°C

Description

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analyte

Description

Product #

Add to Cart

Benzene    
Bromodichloromethane    
Bromoform    
1,3-Dichlorobenzene    
1,4-Dichlorobenzene    
1,2-Dichloroethane    
trans-1,2-Dichloroethylene    
cis-1,3-Dichloropropene    
trans-1,3-Dichloropropene    
Ethylbenzene    
1,1,2,2-Tetrachloroethane    
Toluene    
1,1,1-Trichloroethane    
Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
3
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?