• USA Home
  • 48135 - MISA Group 19 Base-Neutral Extractables Mix B

EMAIL THIS PAGE TO A FRIEND
48135 Supelco

MISA Group 19 Base-Neutral Extractables Mix B

2000 μg/mL each component in methylene chloride, analytical standard

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments, Canada,
grade   analytical standard
feature   standard type calibration
packaging   ampule of 1 mL
concentration   2000 μg/mL each component in methylene chloride
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Agency/Method   MISA (Canada) Group 19
Featured Industry   Environmental
format   multi-component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

Product #

Add to Cart

Benzyl butyl phthalate    
Biphenyl    
Bis(2-ethylhexyl) phthalate    
Dibutyl phthalate    
2,4-Dinitrotoluene    
2,6-Dinitrotoluene    
Di-n-octyl phthalate    
Diphenyl ether    
Indole    
N-Nitrosodiphenylamine    
N-Nitrosodi-n-propylamine    
Perylene    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?