• USA Home
  • 48198 - Heptachlor expoxide isomer A solution

EMAIL THIS PAGE TO A FRIEND
48198 Supelco

Heptachlor expoxide isomer A solution

1000 μg/mL in methanol, analytical standard

Synonym: Heptachlor endo-epoxide solution

  • CAS Number 28044-83-9

  • Empirical Formula (Hill Notation) C10H5Cl7O

  • Molecular Weight 389.32

  •  Beilstein Registry Number 8068917

  •  MDL number MFCD00872082

  •  PubChem Substance ID 24871803

Purchase

Properties

grade   analytical standard
InChI Key   ZXFXBSWRVIQKOD-BDMWXGRVSA-N
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 mL
concentration   1000 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

CRM48198 Heptachlor epoxide Isomer A solution, certified reference material, 1000 μg/mL in methanol, ampule of 1 mL

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?