• USA Home
  • 48265-U - ASTM® D5134 Qualitative Reference Naphtha Standard

EMAIL THIS PAGE TO A FRIEND
48265-U Supelco

ASTM® D5134 Qualitative Reference Naphtha Standard

analytical standard

Purchase

Properties

Related Categories ASTM D5134 Qualitative Reference Materials, ASTM Petroleum Standards, Analytical Standards, Analytical/Chromatography, Fuels and Hydrocarbons,
grade   analytical standard
packaging   pkg of 6 × 1 mL
concentration   (Approx. 100 identified components. Neat fraction.)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Agency/Method   ASTM® D5134
Featured Industry   Petroleum
format   multi-component solution
storage temp.   room temp

Description

Features and Benefits

Approximately 100 identified components. Neat fraction. Actual refinery naphtha product.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993A 3 / PGI
WGK Germany 
3
Flash Point(F) 
69.8 °F
Flash Point(C) 
21 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?