• USA Home
  • 48386 - Pentachlorobenzene solution

EMAIL THIS PAGE TO A FRIEND
48386 Supelco

Pentachlorobenzene solution

certified reference material, 200 μg/mL in methylene chloride

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments,
grade   certified reference material
  TraceCERT®
packaging   ampule of 1 mL
concentration   200 μg/mL in methylene chloride
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
format   single component solution

Description

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Blood, Central nervous system, Liver
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles

Articles

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity®-5 (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method TO-10/IP-8 and ASTM® D4861: GC Analysis of Pesticides on SPB®-608 after Collection/Desorption using ORBO™-1000

From our library of Articles, Sigma-Aldrich presents US EPA Method TO-10/IP-8 and ASTM® D4861: GC Analysis of Pesticides on SPB®-608 after Collection/Desorption using ORBO™-1000
Keywords: Air Monitoring, Chromatography, Electron capture detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?