EMAIL THIS PAGE TO A FRIEND
48592 Supelco

Cyanazine solution

2000 μg/mL in methanol, analytical standard

Purchase

Properties

grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 mL
concentration   2000 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Agency/Method   EPA 508.1
  EPA 508
  EPA 508A
  EPA 629
Featured Industry   Agriculture
Environmental
format   single component solution

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1230 3/PG 2
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?