• USA Home
  • 48661 - Phenanthrene solution

EMAIL THIS PAGE TO A FRIEND
48661 Supelco

Phenanthrene solution

certified reference material, 200 μg/mL in dichloromethane

Purchase

Properties

Related Categories Additional Standards, All Polycyclic Aromatic Hydrocarbon (PAH) Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography,
grade   certified reference material
InChI Key   YNPNZTXNASCQKK-UHFFFAOYSA-N
grade   TraceCERT®
packaging   ampule of 1 mL
concentration   200 μg/mL in dichloromethane
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
format   single component solution
storage temp.   room temp
Gene Information   human ... CYP1A2(1544), CYP2A6(1548)
mouse ... Cyp2a5(13087)

Description

General description

Phenanthrene belongs to the category of polycyclic aromatic hydrocarbons, which can be obtained from various sources like the incomplete combustion of petroleum fuel. It can undergo biodegradation in the presence of A.calcoaceticus and suitable bio-surfactants like non-encapsulated rhamnolipid and herbal surfactants.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

EN Method 2005/108/EC: GC Analysis of Pesticide/PAH Compounds on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents EN Method 2005/108/EC: GC Analysis of Pesticide/PAH Compounds on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Pesticides, Purification

GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Salmon on SPB®-608 (20 m x 0.18 mm I.D., 0.18 μm) after QuEChERS Cleanup using Supel™ QuE Z-Sep, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Salmon on SPB®-608 (20 m x 0.18 mm I.D., 0.18 μm) after QuEChERS Cleanup using Supel™ QuE Z-Sep, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Purification, Sample preparations, Solid phase extractions

GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Soybean Oil on SLB®-35ms after SPE using Supelclean™ EZ-POP NP

From our library of Articles, Sigma-Aldrich presents GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Soybean Oil on SLB®-35ms after SPE using Supelclean™ EZ-POP NP
Keywords: Automotive, Chromatography, Combustion, Environmental, Gas chromatography, Mass selective detector, Mass spectrometry, Sample preparations, Solid phase extractions

GC Analysis of Semivolatiles in Drinking Water on a 5% Phenyl Column after SPE using ENVI™-18 DSK

From our library of Articles, Sigma-Aldrich presents GC Analysis of Semivolatiles in Drinking Water on a 5% Phenyl Column after SPE using ENVI™-18 DSK
Keywords: Chromatography, Gas chromatography, Mass spectrometry, Purification, Sample preparations, Solid phase extractions

GC Analysis of Semivolatiles in Water on a 5% Phenyl Column after SPME using 7 μm PDMS

From our library of Articles, Sigma-Aldrich presents GC Analysis of Semivolatiles in Water on a 5% Phenyl Column after SPME using 7 μm PDMS
Keywords: Chromatography, Environmental, Gas chromatography, Mass spectrometry, Purification, Size-exclusion chromatography, Solid phase microextractions

GC Analysis of a 17-Component Polynuclear Aromatic Hydrocarbon (PAH) Mix on SLB®-IL59

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 17-Component Polynuclear Aromatic Hydrocarbon (PAH) Mix on SLB®-IL59
Keywords: Chromatography, Gas chromatography, Mass selective detector

HPLC Analysis of PAHs in Water on SUPELCOSIL™ LC-PAH after SPME using 100 μm PDMS Fiber

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of PAHs in Water on SUPELCOSIL™ LC-PAH after SPME using 100 μm PDMS Fiber
Keywords: Chromatography, Environmental, Environmental Analysis, High performance liquid chromatography, Solid phase microextractions

HPLC Analysis of PAHs on SUPELCOSIL™ LC-PAH

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of PAHs on SUPELCOSIL™ LC-PAH
Keywords: Chromatography, Environmental, Environmental Analysis, High performance liquid chromatography, Separation

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 610: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 610: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, High performance liquid chromatography, Purification, Separation

US EPA Method 610: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SPB®-608

From our library of Articles, Sigma-Aldrich presents US EPA Method 610: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SPB®-608
Keywords: Chromatography, Gas chromatography, High performance liquid chromatography, Purification, Separation

US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SPB®-608

From our library of Articles, Sigma-Aldrich presents US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SPB®-608
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.
Keywords: Chromatography, Gas chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method TO-13: GC Analysis of PAHs on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method TO-13: GC Analysis of PAHs on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?