• USA Home
  • 48687 - Endosulfan sulfate solution

EMAIL THIS PAGE TO A FRIEND
48687 Supelco

Endosulfan sulfate solution

20 μg/mL in methanol, analytical standard

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Cyclodienes, E-G,
grade   analytical standard
InChI Key   AAPVQEMYVNZIOO-WINLOITPSA-N
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 mL
concentration   20 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   single component solution
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of Agricultural Pesticides (Standard) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of Agricultural Pesticides (Standard) on SLB®-5ms
Keywords: Agriculture, Chromatography, Gas chromatography, Gas chromatography mass spectrometry, Herbicides, Mass spectrometry, Pesticides

GC Analysis of Agriculture Pesticides in Orange (Spiked) on SLB®-5ms after QuEChERS Cleanup using Primary-Secondary Amine (PSA)

From our library of Articles, Sigma-Aldrich presents GC Analysis of Agriculture Pesticides in Orange (Spiked) on SLB®-5ms after QuEChERS Cleanup using Primary-Secondary Amine (PSA)
Keywords: Chromatography, Gas chromatography, Pesticides, Purification, Sample preparations, Solid phase extractions

GC Analysis of Multiple Pesticides on SLB®-5ms after SPME using 60 μm Carbowax® Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Multiple Pesticides on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
Keywords: Agriculture, Chromatography, Gas chromatography, Metabolites, Pesticides, Purification, Solid phase microextractions

GC Analysis of Organochlorine Pesticides in Water on SPB®-5 after SPME using 100 μm PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Organochlorine Pesticides in Water on SPB®-5 after SPME using 100 μm PDMS Fiber
Keywords: Chromatography, Electron capture detector, Environmental, Gas chromatography, Metabolites, Pesticides, Purification, Size-exclusion chromatography, Solid phase microextractions

GC Analysis of Pesticides in Beef Kidney on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm) after QuEChERS Cleanup using Supel™ Que Z-Sep+, MS in SIM Mode, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Pesticides in Beef Kidney on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm) after QuEChERS Cleanup using Supel™ Que Z-Sep+, MS in SIM Mode, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Pesticides, Purification, Sample preparations, Solid phase extractions

GC Analysis of a 20-Component Organochlorine Pesticide Mix in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI™-8

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 20-Component Organochlorine Pesticide Mix in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI™-8
Keywords: Chromatography, Electron capture detector, Gas chromatography, Purification, Sample preparations, Size-exclusion chromatography, Solid phase extractions

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on Equity®-1701 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on Equity®-1701 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Electron capture detector, Gas chromatography, Pesticides, Purification, Size-exclusion chromatography

US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Electron capture detector, Gas chromatography, Pesticides, Purification, Size-exclusion chromatography

US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SLB®-5ms
Keywords: Chromatography, Electron capture detector, Gas chromatography, Pesticides, Purification

US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SPB®-608

From our library of Articles, Sigma-Aldrich presents US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on SPB®-608
Keywords: Chromatography, Electron capture detector, Gas chromatography, Pesticides, Purification

US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8081: GC Analysis of Organochlorine Pesticides on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8081: GC Analysis of Organochlorine Pesticides on Equity®-5
Keywords: Chromatography, Electron capture detector, Environmental, Gas chromatography, Pesticides, Purification, Size-exclusion chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?