• USA Home
  • 48702 - Aroclor 1232 solution

EMAIL THIS PAGE TO A FRIEND
48702 Supelco

Aroclor 1232 solution

certified reference material, 200 μg/mL in methanol

Purchase

Properties

Related Categories 500 Series Drinking Water Methods, 8000 Series Solidwaste Methods, Additional Standards, Analytical Standards, Analytical/Chromatography,
grade   certified reference material
  TraceCERT®
feature   standard type calibration
packaging   ampule of 1 mL
concentration   200 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Agency/Method   EPA 505
  EPA 508
  EPA 8082
  MISA (Canada) Group 27
Featured Industry   Environmental
format   single component solution
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

Method 508A - Determination of Chlorinated Pesticides in Water

Products listed below are for use in EPA Method 508A: "Screening for Polychlorinated Biphenyls by Perchlorination and Gas Chromatography."
Keywords: Chromatography, Gas chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?