• USA Home
  • 48805-U - EPA Phthalate Esters Mix

EMAIL THIS PAGE TO A FRIEND
48805-U Supelco

EPA Phthalate Esters Mix

2000 μg/mL each component in methanol, analytical standard

Purchase

Properties

Related Categories 500 Series Drinking Water Methods, 600 Series Wastewater Methods, 8000 Series Solidwaste Methods, Additional Standards, Analytical Standards,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 mL
concentration   2000 μg/mL each component in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   multi-component solution

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

Bis(2-ethylhexyl) phthalate SDS    
Benzyl butyl phthalate SDS    
Dibutyl phthalate SDS    
Di-n-octyl phthalate SDS    
Diethyl phthalate SDS    
Dimethyl phthalate SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1230 3/PG 2
WGK Germany 
2
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

CRM48805 EPA Phthalate Esters Mix, certified reference material, 2000 μg/mL each component in methanol, ampule of 1 mL

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?