EMAIL THIS PAGE TO A FRIEND
48861 Supelco

Aroclor Mix 1

certified reference material, 200 μg/mL each component in methanol

Purchase

Properties

Related Categories 600 Series Wastewater Methods, ASTM Air Monitoring, ASTM D4861: Analysis of PCBS in Air, Additional Standards, Analytical Standards,
Quality Level   100
grade   certified reference material
Agency/Method   ASTM® D4861
  EPA 608,625
  NIOSH 5503
grade   TraceCERT®
feature   standard type calibration
packaging   ampule of 1 mL
concentration   200 μg/mL each component in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
format   multi-component solution
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

Product #

Add to Cart

Aroclor 1016    
Aroclor 1232    
Aroclor 1248    
Aroclor 1260    
Safety & Documentation

Safety Information

Signal word 
Danger
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?