• USA Home
  • 48864 - EPA Purgeable Internal Standard Mix

EMAIL THIS PAGE TO A FRIEND
48864 Supelco

EPA Purgeable Internal Standard Mix

certified reference material, 20,000 μg/mL each component in methanol

Purchase

Properties

Quality Level   100
grade   certified reference material
Agency/Method   EPA 601,624
grade   TraceCERT®
feature   standard type internal
packaging   ampule of 1 mL
concentration   20,000 μg/mL each component in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Environmental
format   multi-component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

Product #

Add to Cart

Bromochloromethane    
2-Bromo-1-chloropropane    
1,4-Dichlorobutane    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?