EMAIL THIS PAGE TO A FRIEND
48866 Supelco

EPA 625 Phenol Mix

certified reference material, in dichloromethane (varied)

Purchase

Properties

Related Categories 600 Series Wastewater Methods, Additional Standards, Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments,
grade   certified reference material
  TraceCERT®
feature   standard type calibration
packaging   ampule of 1 mL
concentration   in dichloromethane (varied)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Agency/Method   EPA 625
Featured Industry   Environmental
format   multi-component solution
storage temp.   room temp

Description

Application

EPA 625 phenol mix, which can be used for extractable bases/neutrals and acids, can find applications in the determination of phenols using dichloromethane as the extraction solvent via gas chromatography coupled with mass spectrometry method (GC-MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

TThis Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

Product #

Add to Cart

4-Chloro-3-methylphenol 2500 μg/mL    
2-Chlorophenol 500 μg/mL    
2,4-Dichlorophenol 500 μg/mL    
2,4-Dimethylphenol 500 μg/mL    
2,4-Dinitrophenol 2500 μg/mL    
2-Methyl-4,6-dinitrophenol 2500 μg/mL    
2-Nitrophenol 500 μg/mL    
4-Nitrophenol 2500 μg/mL    
Pentachlorophenol 2500 μg/mL    
Phenol 500 μg/mL    
2,4,6-Trichlorophenol 1500 μg/mL    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?