EMAIL THIS PAGE TO A FRIEND
48930-U Supelco

ASTM® PS-12-60N

analytical standard

Purchase

Properties

Related Categories ASTM Petroleum and Petrochemical, Additional Standards, Analytical Standards, Analytical/Chromatography, Fuels and Hydrocarbons,
grade   analytical standard
form   neat
packaging   pkg of 500 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Petroleum
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

Product #

Add to Cart

Docosane 6.9 % (w/w)    
Dodecane 1.72 % (w/w)    
Dotriacontane 6.9 % (w/w)    
Eicosane 5.17 % (w/w)    
Hexacontane 1.72 % (w/w)    
Hexacosane 17.24 % (w/w)    
Hexadecane 3.45 % (w/w)    
Hexatriacontane 5.17 % (w/w)    
Octacosane 17.24 % (w/w)    
Octadecane 4.31 % (w/w)    
Pentacontane 2.59 % (w/w)    
Tetracontane 4.31 % (w/w)    
Tetracosane 8.62 % (w/w)    
Tetradecane 2.5 % (w/w)    
Tetratetracontane 3.5 % (w/w)    
Triacontane 8.62 % (w/w)    
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?