EMAIL THIS PAGE TO A FRIEND
49017 Supelco

4,4′-DDE

analytical standard, ampule of 1000 mg

Synonym: 1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene, 4,4′-DDE

  • CAS Number 72-55-9

  • Linear Formula (ClC6H4)2C=CCl2

  • Molecular Weight 318.03

  •  Beilstein Registry Number 1913355

  •  EC Number 200-784-6

  •  MDL number MFCD00000837

  •  PubChem Substance ID 24872533

Purchase

Properties

grade   analytical standard
InChI Key   UCNVFOCBFJOQAL-UHFFFAOYSA-N
packaging   ampule of 1000 mg
mp   88-90 °C (lit.)
Gene Information   mouse ... Esr1(13982)
rat ... Ar(24208)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
KV9450000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?