EMAIL THIS PAGE TO A FRIEND
49086 Supelco

Prometon

analytical standard

Synonym: 2,4-Bis-(isopropylamino)-6-methoxy-1,3,5-triazine

  • CAS Number 1610-18-0

  • Empirical Formula (Hill Notation) C10H19N5O

  • Molecular Weight 225.29

  •  Beilstein/REAXYS Number 613574

  •  EC Number 216-548-0

  •  MDL number MFCD00047343

  •  PubChem Substance ID 24872610

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 100 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   neat
storage temp.   room temp
SMILES string   COc1nc(NC(C)C)nc(NC(C)C)n1
InChI   1S/C10H19N5O/c1-6(2)11-8-13-9(12-7(3)4)15-10(14-8)16-5/h6-7H,1-5H3,(H2,11,12,13,14,15)
InChI key   ISEUFVQQFVOBCY-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
XY4200000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

GC Analysis of Multiple Pesticides on SLB®-5ms after SPME using 60 μm Carbowax® Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Multiple Pesticides on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
Keywords: Agriculture, Chromatography, Gas chromatography, Metabolites, Pesticides, Purification, Solid phase microextractions

GC Analysis of Triazine Herbicides on SLB®-5ms after SPME using 60 μm Carbowax® Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Triazine Herbicides on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
Keywords: Agriculture, Chromatography, Gas chromatography, Herbicides, Purification, Solid phase microextractions

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?