EMAIL THIS PAGE TO A FRIEND
4S8777 Supelco

EPA VOC Mix 2

Separate Source, certified reference material, 2000 μg/mL each component in methanol

Purchase

Properties

Related Categories 500 Series Drinking Water Methods, 8000 Series Solidwaste Methods, Additional Standards, Alpha Sort, Analytical Standards,
grade   certified reference material
description   Separate Source
grade   TraceCERT®
feature   standard type calibration
packaging   ampule of 1 × 1 mL (4S8777)
concentration   2000 μg/mL each component in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Agency/Method   EPA 502.1
  EPA 502.2
  EPA 503.1
  EPA 524.1
  EPA 524.2
  EPA 8021 (1987)
  EPA 8021A (1990)
  EPA 8260
Featured Industry   Environmental
format   multi-component solution
storage temp.   2-8°C

Description

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Footnote

Separate Source standard. Prepared from independently sourced raw material and quality controlled independently.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

Product #

Add to Cart

Benzene    
Bromobenzene    
Butylbenzene    
Ethylbenzene    
Naphthalene    
Styrene    
Toluene    
1,2,3-Trichlorobenzene    
1,2,4-Trichlorobenzene    
1,2,4-Trimethylbenzene    
m-Xylene    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
3
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?