• USA Home
  • 502669 - Upchurch Precolumn MicroFilter

502669 Supelco

Upchurch Precolumn MicroFilter

for use with 0.025 in. O.D. tubing, stainless steel frit



Related Categories Analytical/Chromatography, HPLC, HPLC Accessories, Upchurch Precolumn MicroFilter, Upchurch Precolumn MicroFilter and Frits,
material   PEEK body
  stainless steel frit
packaging   pkg of 1 ea
fitting   10-32 female Upchurch for tubing 0.025 in. O.D. requires MicroTight tubing sleeve (nut included)
  10-32 male Upchurch 1/16 in.
compatibility   for use with 0.025 in. O.D. tubing


General description

PEEK housing with in-line filter connects capillary tubing (using Microtight tubing sleeves) or 1/16 in. O.D. tubing to female 10-32 fitting. Swept volume of 0.3 μL. Includes 5 × 0.5 μm frits (one installed).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Price and Availability

Milli-Q® Water Purification Solutions
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.


Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?