• USA Home
  • 506427 - ASTM® D3710 Qualitative Calibration Mix

EMAIL THIS PAGE TO A FRIEND
506427 Supelco

ASTM® D3710 Qualitative Calibration Mix

analytical standard

Purchase

Properties

Related Categories ASTM Petroleum and Petrochemical, Additional Standards, Analytical Standards, Analytical/Chromatography, Fuels and Hydrocarbons,
grade   analytical standard
packaging   pkg of 1 mL
concentration   (ach component in the approximate proportions (w/w) indicated)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Agency/Method   ASTM® D3710
Featured Industry   Petroleum
format   multi-component solution
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

Product #

Add to Cart

Butane 4.5 % (w/w)    
Butylbenzene 3.2 % (w/w)    
Decane 3.2 % (w/w)    
2,4-Dimethylpentane 5.4 % (w/w)    
Dodecane 3.2 % (w/w)    
Heptane 9.7 % (w/w)    
Hexane 5.4 wt. %    
2-Methylbutane 9.7 % (w/w)    
2-Methylpentane 5.4 % (w/w)    
2-Methylpropane 1.5 % (w/w)    
Octane 5.4 % (w/w)    
Pentadecane 2.2 % (w/w)    
Pentane 7.6 % (w/w)    
Propane 1.5 % (w/w)    
Propylbenzene 4.3 % (w/w)    
Tetradecane 2.2 % (w/w)    
Toluene 10.8 % (w/w)    
Tridecane 2.2 % (w/w)    
p-Xylene 13 wt. %    
Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN3295 - class 3 - PG 2 - EHS - VP > 110kPa - Hydrocarbons, liquid, n.o.s., HI: A, S (not BR)
WGK Germany 
3
Flash Point(F) 
-99.4 °F
Flash Point(C) 
-73 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?