• USA Home
  • 50687-U - Hamilton PRP-C-18 Preparative Guard Column Starter Kit

EMAIL THIS PAGE TO A FRIEND
50687-U

Hamilton PRP-C-18 Preparative Guard Column Starter Kit

12-20 μm particle size, L × I.D. 20 mm × 4.6 mm, stainless steel, kit of

  •  NACRES SB.52

Purchase

Properties

Related Categories Analytical/Chromatography, Established Column Lines from Other Manufacturers, HPLC Columns, by Brand for Small Molecules, HPLC/UHPLC, Hamilton HPLC Columns,
material   stainless steel
mfr. no.   Hamilton
packaging   kit of
application(s)   HPLC: suitable
L × I.D.   20 mm × 4.6 mm
matrix   PS-DVB particle platform
matrix active group   C18 (octadecyl) phase
particle size   12-20 μm
pore size   100 Å
separation technique   reversed phase

Description

General description

Kit contains 1 holder, 1 cartridge

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?