• USA Home
  • 507733 - Septa, PTFE/red rubber, pkg 100
EMAIL THIS PAGE TO A FRIEND
507733 Supelco
Septa, PTFE/red rubber, pkg 100
PTFE/red rubber (38 durometer), diam. 8 mm
Purchase
Properties
Related Categories Analytical/Chromatography, Chromacol Septa for 2 mL vial, 8 mm diameter, Screw Top Autosampler Vials, 1.5 to 4 mL, Screw Top and Shell Vials, 12 x 32 mm, 4.6 mm Opening Taking 8 mm Septa, Vials,
material   PTFE septum face
  PTFE/red rubber
feature   temp. limit 120 °C
  PTFE/red rubber (38 durometer)
packaging   pkg of 100 ea
mfr. no.   Chromacol, 8-6RT1
diam.   8 mm
Description
General description
It is used as an air-tight seal, and prevents contamintion
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
27094-U Screw cap, phenolic, with open center, black phenolic hole cap, thread 8-425, no septum, pkg of 100 ea
27401 Screw cap, black polypropylene hole cap, large, opening in top of cap, no septum, thread 8-425, pkg of 100 ea
507601 Vial, Chromacol, volume 2 mL, clear glass vial, O.D. × H × I.D. 12 mm × 32 mm × 4.6 mL/ft, thread 8-425, Chromacol, 2-SV, pkg of 100 ea
507628 Vials, screw top, amber glass (vial only), volume 2 mL, amber glass vial, thread 8-425, pkg of 100 ea
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?