• USA Home
  • 507784 - Septa, white PTFE/silicone

507784 Supelco

Septa, white PTFE/silicone

diam. × thickness 8 mm × 0.060 in., durometer 20, for use with 2 mL vial - standard opening, pkg of 100 ea



Related Categories Analytical/Chromatography, Material, PTFE/Silicone, Sampling Vials, Caps, and Septa, Sampling, Storage, and Serum Bottles,
material   PTFE/silicone
feature   temp. limit 250 °C
  temp. limit 250 °C
packaging   pkg of 100 ea
diam. × thickness   8 mm × 0.060 in.
compatibility   for use with 2 mL vial - standard opening

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.


Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #


Add to Cart

27094-U Screw cap, phenolic, with open center, black phenolic hole cap, thread 8-425, no septum, pkg of 100 ea
27401 Screw cap, black polypropylene hole cap, large, opening in top of cap, no septum, thread 8-425, pkg of 100 ea
507601 Vial, Chromacol, volume 2 mL, clear glass vial, O.D. × H × I.D. 12 mm × 32 mm × 4.6 mL/ft, thread 8-425, Chromacol, 2-SV, pkg of 100 ea
507628 Vials, screw top, amber glass (vial only), volume 2 mL, amber glass vial, thread 8-425, pkg of 100 ea
507660 Screw cap (open-top), polypropylene, white polypropylene hole cap, size 8-425, pkg of 100 ea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?