• USA Home
  • 51105AST - Astec® Cellulose DMP Chiral HPLC Guard

EMAIL THIS PAGE TO A FRIEND
51105AST Supelco

Astec® Cellulose DMP Chiral HPLC Guard

Kit, 5 μm particle size, L × I.D. 2 cm × 2.1 mm

  •  eCl@ss 32110501

  •  NACRES SB.52

Purchase

Properties

Related Categories Analytical/Chromatography, Astec Cellulose DMP Chiral HPLC Columns (Polysaccharide-based), Chiral HPLC Columns, Chiral HPLC Guard Columns and Accessories, HPLC/UHPLC More...
Quality Level   100
packaging   pkg of 1 kit
application(s)   HPLC: suitable
L × I.D.   2 cm × 2.1 mm
matrix   silica gel particle platform
matrix active group   cellulose, DMPC- phase
particle size   5 μm
suitability   suitable for L40 per USP
separation technique   chiral

Description

General description

Guard cartridges to protect Astec® Cellulose DMP columns. Choose the I.D. to match the I.D. of your analytical column. The 2.1 mm and 4 mm I.D. cartridges are sold in kits that contain the holder and fittings, and as packs of 2 cartridges. The 10 mm and 21.2 mm I.D. guards are sold as individual cartridges that require holders, sold separately.

Guard column kit. Contains holder, one 2 cm x 2.1 mm I.D. Astec® Cellulose DMP cartridge (5 μm particles), and 1/16" x 0.010" I.D. tubing (5 cm), two nuts and two ferrules.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?