• USA Home
  • 51173-U - OPTI-SOLV® Mini, Micro, and Nano Filter
EMAIL THIS PAGE TO A FRIEND
51173-U
OPTI-SOLV® Mini, Micro, and Nano Filter
micro filter, 2.0μm, pkg of 5 ea
  •  NACRES SB.52
Purchase
Description
General description
The OPTI-SOLV® Mini Filter provides low-impact filtering in a package no longer that a finger tight fitting. Use them to prolong the life of your analytical colum, or before your mass spectrometer as a last line of defense against debris.
The OPTI-SOLV® Micro Filter is based on the same design as the Mini Filter and cuts the internal volume to less that 200 nL. It retains the Mini Filter′s ease of use and functionality. The OPTI-SOLV® Micro Filter features a zero-dead volume connection utilizing Optimize Technologies® patented floating stem technology.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
OPTI-SOLV is a registered trademark of Optimize Technologies, Inc.
Optimize Technologies is a registered trademark of Optimize Technologies, Inc.
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?