• USA Home
  • 51196-U - OPTI-GUARD® 3 mm Guard Column

EMAIL THIS PAGE TO A FRIEND
51196-U

OPTI-GUARD® 3 mm Guard Column

Silica cartridge, pkg of 3 ea

  •  eCl@ss 32110501

  •  NACRES SB.52

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC Accessories, HPLC Columns, by Brand for Small Molecules, HPLC/UHPLC, OPTI-GUARD 3mm Guard Column,
packaging   pkg of 3 ea
mfr. no.   OPTI-GUARD
matrix active group   silica phase
particle size   5 μm
separation technique   normal phase

Description

General description

The OPTI-GUARD® 3 mm maintains the tool free connectivity of the OPTI-GUARD® 1 mm, but incorporates a cartridge-based format to allow for a larger bed with more capacity.

The two part holder is designed for use with any analytical column providing vital protection from dirty samples and strongly retained contaminants. An auto adjusting stem conforms perfectly to any tube stop depth resulting in a perfect zero-volume connection.

Note: cartridges not included with holder.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

OPTI-GUARD is a registered trademark of Optimize Technologies, Inc.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Frequently Asked Questions

Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
How do I get lot-specific information or a Certificate of Analysis?
The lot specific COA document can be found by entering the lot number above under the "Documents" section.
How do I find price and availability?
There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote. USA customers: 1-800-325-3010 or view local office numbers.
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
What is the pore size and particle size of the material in Opti-Guard® HPLC Guard Columns?
The pore size is 100 Angstroms and the particle size is 40μm for the Opti-Guard® HPLC Guard Columns.
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.
Show more questions
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?