• USA Home
  • 52134AST - Astec® C18 HPLC Guard Column

52134AST Supelco

Astec® C18 HPLC Guard Column

5 μm particle size, L × I.D. 2 cm × 4 mm

  •  eCl@ss 32110501



Related Categories Analytical/Chromatography, Astec Silica-based, C18 Reversed Phase, HPLC, HPLC Columns, by Brand for Small Molecules More...
packaging   pkg of 1 ea
application(s)   HPLC: suitable
L × I.D.   2 cm × 4 mm
matrix active group   C18 (octadecyl) phase
particle size   5 μm
pore size   110 Å
suitability   suitable for L1 per USP
mode of chromatography   reversed phase


Legal Information

Astec is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Milli-Q® Water Purification Solutions
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.


Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?