• USA Home
  • 52233-U - Upchurch VHP (Very High Pressure) Tools
EMAIL THIS PAGE TO A FRIEND
52233-U
Upchurch VHP (Very High Pressure) Tools
VHP-4000 Adustable torque tightening screwdriver
  •  NACRES SB.52
Purchase
Properties
Related Categories Analytical/Chromatography, HPLC Accessories, HPLC/UHPLC, Upchurch Scientific Fittings and Accessories, Upchurch VHP (Very High Pressure) Fittings,
description   2 to 12 inch lbs. with 1/4 inch female socket
mfr. no.   VHP-4000 Adustable torque tightening screwdriver
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?