• USA Home
  • 52734-U - SupelMIP® SPE - Nitroimidazoles

EMAIL THIS PAGE TO A FRIEND
52734-U Supelco

SupelMIP® SPE - Nitroimidazoles

bed wt. 50 mg, volume 3 mL, pk of 50

Purchase

Properties

Related Categories Analytical/Chromatography, Sample Preparation & Purification, Solid Phase Extraction (SPE), SupelMIP Molecular Imprinted Polymers, SupelMIP SPE - Nitroimidazoles More...
composition   bed wt., 50 mg
packaging   pk of 50
volume   3 mL
matrix active group   SupelMIP® phase

Description

Application

Developed and optimized for the extraction of nitroimidazoles from a variety of sample matrixes including milk and eggs.

Example nitroimidazoles include dimetridazole (DMZ), ipronidazole (IPZ), metronidazole (MNZ), and ronidazole (RNZ) and their hydroxylated metabolites (DMZOH and MNZOH)

Features and Benefits

• Highly selective SPE for trace analysis in complex matrices
• Achieve lower detection limits
• Significant time and cost savings
• Improved MS-compatibility

General description

SupelMIP SPE phases were developed by MIP Technologies AB, which is one of the leading authorities and commercial pioneers of molecular imprinted polymers for process scale separations, analytical chromatography, and sample preparation.

The SupelMIP SPE line consist s of highly cross-linked polymers that are engineered to extract a single analyte of interest or a class of structurally related analytes with an extremely high degree of selectivity. This is possible because selectivity is introduced during MIP synthesis in which a template molecule, designed to mimic the analyte, guides the formation of specific cavities or imprints that are sterically and chemically complementary to the analyte(s) of interest.

By careful design of the imprinting site, either by molecular modeling, experimental design, or screening methods, the binding cavities can be engineered to offer multiple interaction points with the analyte(s) of interest. This leads to a stronger interaction between the solid-phase and the analyte(s). As a consequence, harsher wash conditions can be tolerated during SPE methodology resulting in cleaner extracts. Because extraction selectivity is significantly improved, lower background is observed allowing analysts to achieve lower limits of detection.

Visit www.sigma-aldrich.com/supelmip to learn more.

Legal Information

SupelMIP is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles

Related Content

SPE Cartridges (Tubes) - Phase Selection Guide

The SPE Tube Configuraton Guide will help you choose the appropriate bed weight and cartridge dimensions.
Keywords: Agriculture, Calorimetry, Environmental, Fractionation, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Pesticides, Pharmaceutical, Solid phase extractions

Solid Phase Extraction: Fundamentals and Recent Developments [Webinar]

Solid Phase Extraction (SPE) has become a popular technique for sample preparation due to its convenience, effectiveness, relatively low solvent usage and versatility. The focus of this webinar is on...
Keywords: Sample preparations, Solid phase extractions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?