• USA Home
  • 52806-U - HPLC Dispersion Measurement Kit
EMAIL THIS PAGE TO A FRIEND
52806-U Supelco
HPLC Dispersion Measurement Kit
Purchase
Description
General description
Kit for reliable measurement of HPLC instrument bandwidth.
Kit contains:
2 - NanoTight Unions with 0.007" thru-hole
1 - 20 cm x 1/32" O. D. x 0.005" I. D. High Performance Doubled Ended Fitting
Instruction Sheet
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?