• USA Home
  • 54262-U - Ascentis® Express C8 Capillary, 2.7 micron HPLC Column

EMAIL THIS PAGE TO A FRIEND
54262-U Supelco

Ascentis® Express C8 Capillary, 2.7 micron HPLC Column

2.7 μm particle size, L × I.D. 15 cm × 200 μm

  •  eCl@ss 32110501

Purchase

Properties

Related Categories Analytical/Chromatography, Ascentis Express C8 Columns, Ascentis Express C8, 2.7μm HPLC Columns, Ascentis Express Columns for Fast HPLC, C8, 2.7μm Capillary Columns,
feature   endcapped
parameter   60 °C temp. range
application(s)   HPLC: suitable
L × I.D.   15 cm × 200 μm
impurities   <5 ppm metals
matrix   Fused-Core particle platform
matrix active group   C8 (octyl) phase
particle size   2.7 μm
pore size   90 Å
operating pH range   2 - 9
suitability   suitable for L7 per USP
separation technique   reversed phase

Description

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Ascentis is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Frequently Asked Questions

Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
How do I get lot-specific information or a Certificate of Analysis?
The lot specific COA document can be found by entering the lot number above under the "Documents" section.
What is special about Ascentis Express?
Ascentis Express columns provide a breakthrough in HPLC performance. Based on Fused-Core particle technology, Ascentis Express provides the benefits of sub-2 μm particles but at much lower backpressure.These benefits include the capability of providing fast HPLC and higher resolution chromatography.The Fused-Core particle consists of a 1.7 μm solid core and a 0.5 μm porous shell.A major benefit of the Fused-Core particle is the small diffusion path (0.5 μm) compared to conventional fully porous particles. The shorter diffusion path reduces axial dispersion of solutes and minimizes peak broadening.
When are additional phases expected?
Additional phases are being developed.The best way to track new products is to visitAscentis Express for the latest updates.
Can I use Ascentis Express on any type of HPLC system?
Ascentis Express HPLC columns are capable of use on standard HPLC systems as well as UHPLC systems.Columns are packed in high pressure hardware capable of withstanding the pressures used in UHPLC systems.
Can I use Ascentis Express on a UHPLC system?
Yes.Ascentis Express columns are packed in a way making them suitable for these ultra high pressure instruments.In fact, Ascentis Express outperforms sub-2 μm micron columns on many applications since Ascentis Express provides the benefits of sub-2 μm particles but at much lower back pressure.These benefits include the capability of providing fast HPLC and higher resolution chromatography.The Fused-Core particle consists of a 1.7 μm solid core and a 0.5 μm porous shell.A major benefit of the Fused-Core particle is the small diffusion path (0.5 μm) compared to conventional fully porous particles. The shorter diffusion path reduces axial dispersion of solutes and minimizes peak broadening.
How do I find price and availability?
There are several ways to find pricing and availability for our products.Once you log onto our website, you will find the price and availability displayed on the product detail page.You can contact any of our Customer Sales and Service offices to receive a quote. USA customers: 1-800-325-3010 orview local office numbers.
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
What is the pressure rating for 200 μm I.D. Ascentis® Express Capillary HPLC columns?
The 200 μm I.D. Ascentis® Express Capillary HPLC columns are stable to operating pressures up to 6500 psi.
What is the carbon load for Ascentis Express C8?
Carbon load which is commonly referred to as %Carbon is misleading and not meaningful for Ascentis Express. Since Fused-Core particle is unique in the sense that it has a solid core and porous shell, the %Carbon number can not be compared to traditional porous particles.The carbon coverage for Ascentis Express C8 is 3.7 micromoles/meter-squared
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.
Show more questions

Milli-Q® Water Purification Solutions
Protocols & Articles

Articles

Ascentis Express UHPLC Columns with 2.0 μm Fused-Core Particles

The new Supelco Ascentis® Express column line has been designed specifically for UHPLC instruments with 2.0 µm Fused-Core® silica, as diagrammed in Figure 1. Ascentis Express 2.0 µm UHPLC columns are...
Richard A. Henry, Gaurang Parmar
Reporter US, Volume 32.2
Keywords: High performance liquid chromatography, Mass spectrometry, Pharmaceutical, Separation

Related Content

Selection Guide for Ascentis® Express HPLC Columns

Ascentis Express C18 is the first choice for starting a new method. However, when a C18 doesn't give the desired separation or your sample contains compounds that are known to be difficult to retain ...
Keywords: High performance liquid chromatography, Separation, Supercritical fluid chromatography

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?