• USA Home
  • 54408 - Rheodyne® 3-WAY PTFE ROTARY VALVE 0.8MM BORE (#5031)
EMAIL THIS PAGE TO A FRIEND
54408
Rheodyne® 3-WAY PTFE ROTARY VALVE 0.8MM BORE (#5031)
Purchase
Properties
material   Kel-F housing
  PTFE rotary valve
feature   manual 3-way valve
  number of ports 6
  number of positions 2
mfr. no.   Rheodyne, 5031
parameter   300 psi max. pressure
fitting   1/4-28 female valve ports for 1/16 in. O.D. tubing
Description
Legal Information
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?