• USA Home
  • 54429 - Rheodyne® 6-POSITION SELECT OR VALVE, PEEK (#9060)
EMAIL THIS PAGE TO A FRIEND
54429
Rheodyne® 6-POSITION SELECT OR VALVE, PEEK (#9060)
Purchase
Properties
material   PEEK
feature   manual
  number of ports 7
  number of positions 6
mfr. no.   Rheodyne 9060
parameter   5000 psi max. pressure
fitting   10-32 Rheodyne selector
compatibility   for use with 1/16 in. tubing
Description
Legal Information
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?