• USA Home
  • 54843 - SUPELCOSIL LC-Si HPLC Column

EMAIL THIS PAGE TO A FRIEND
54843 Supelco

SUPELCOSIL LC-Si HPLC Column

5 μm particle size, L × I.D. 25 cm × 21.2 mm

  •  eCl@ss 32110501

Purchase

Properties

Related Categories 5 μm, 5 micron LC-Si silica Columns, Analytical/Chromatography, HPLC, HPLC Columns, by Brand for Small Molecules,
parameter   ≤70 °C temp. limit
application(s)   HPLC: suitable
L × I.D.   25 cm × 21.2 mm
matrix   silica gel, spherical particle platform
matrix active group   silica phase
particle size   5 μm
pore size   120 Å pore size
surface area   170 m2/g
pH-range   2 - 7.5
suitability   suitable for L3 per USP
mode of chromatography   hydrophilic interaction (HILIC)
  normal phase

Description

General description

A typical mobile phase used with normal-phase silica columns consists of a hydrocarbon solvent such as hexane or heptane, mixed with a relatively small percentage of more polar solvent. Non-polar compounds will elute from the column first, while polar solutes show stronger interaction with the silanol groups on the silica surface. The polar selectivity of silica is very helpful for separating mycotoxins. Silica is frequently used in preparative chromatography due to its low operating cost and back pressure compared to reversed-phase columns. Silica is particularly popular among organic chemists as a low-pressure preparative tool. The unmodified silica in SUPELCOSIL LC-Si columns also separate positional isomers.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

58281 Normal Phase Test Mix, in methylene chloride (varied conc.), analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?