• USA Home
  • 55208 - Frit Cap Assembly for Supelco® HPLC Column Hardware

EMAIL THIS PAGE TO A FRIEND
55208 Supelco

Frit Cap Assembly for Supelco® HPLC Column Hardware

size 3.0 mm, pore size 2 μm

Purchase

Properties

Related Categories 3.0 mm, Analytical/Chromatography, Column Hardware, Frit Assemblies for Supelco Column Hardware, HPLC Accessories,
material   PEEK with stainless steel frit
feature   color-coded gray
packaging   pkg of 2 ea
size   3.0 mm
pore size   2 μm
compatibility   for use with Supelco 3.0mm I.D. Discovery and Supelcosil columns
  use to protect 5μm and greater packing material

Description

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Supelco is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?