• USA Home
  • 56039-U - Omnifit® Tubing Cone

EMAIL THIS PAGE TO A FRIEND
56039-U

Omnifit® Tubing Cone

size 1/8  in. , bore

  •  NACRES SB.52

Purchase

Description

Legal Information

Omnifit is a registered trademark of Diba Industries

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?