• USA Home
  • 56106AST - apHera C18 HPLC Column

EMAIL THIS PAGE TO A FRIEND
56106AST Supelco

apHera C18 HPLC Column

5 μm particle size, L × I.D. 25 cm × 6 mm

  •  eCl@ss 32110501

  •  NACRES SB.52

Purchase

Properties

Related Categories Analytical/Chromatography, HPLC Columns, by Brand for Small Molecules, HPLC/UHPLC, apHera C18 Reversed-phase Columns, apHera Polymeric HPLC columns, high pH applications More...
Quality Level   100
packaging   pkg of 1 ea
mfr. no.   apHera
parameter   147 bar pressure (2133 psi)
  4-60 °C temperature
application(s)   HPLC: suitable
L × I.D.   25 cm × 6 mm
matrix active group   C18 (octadecyl) phase
particle size   5 μm
pore size   300 Å
suitability   suitable for L67 per USP
Featured Industry   Food and Beverages
separation technique   reversed phase

Description

General description

apHera reversed columns were devoloped specifically to provide the superior advantages of both silica and polystyrene columns, without the disadvantages of either. This was accomplished using a vinyl alcohol copolymer base that keeps the surface wetted even with high carbon loads. The porous structure has an average pore diameter large enough to produce ideal results for small analytes, peptides and small proteins. These columns equal silica based columns in separation efficiency with organic solvents but provide efficiency with buffered alkaline solutions not possible on silica. One of the most significant features is logicial elution order of alkylated bases where retention increases proportionately with increasing chain length.

Features of apHera Reversed Phase Columns
• Stable vinyl copolymer base
• Stable pH range 2-12
• 300 angstrom pore size
• Stable in all organic solvents
• Expanded applications: peptides, proteins
• Efficient separation of any basic substance
• Amenability to washing and alkaline solutions

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

apHera is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

guard column

Product #

Description

Add to Cart

56131AST apHera C18 HPLC Guard Column, 5 μm particle size, L × I.D. 1 cm × 6 mm

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?