• USA Home
  • 56433AST - apHera NH2 HPLC Guard Column

EMAIL THIS PAGE TO A FRIEND
56433AST Supelco

apHera NH2 HPLC Guard Column

13 μm particle size, L × I.D. 5 cm × 7.5 mm

  •  eCl@ss 32110501

  •  NACRES SB.52

Purchase

Properties

Related Categories Amino HPLC Columns, Analytical/Chromatography, HPLC Columns, by Brand for Small Molecules, HPLC Columns, by Phase, HPLC/UHPLC,
Quality Level   100
packaging   pkg of 1 ea
mfr. no.   apHera
parameter   147 bar pressure (2133 psi)
  4-50 °C temperature
application(s)   HPLC: suitable
L × I.D.   5 cm × 7.5 mm
matrix active group   amino phase
particle size   13 μm
pore size   300 Å
suitability   suitable for L82 per USP
Featured Industry   Food and Beverages
separation technique   hydrophilic interaction (HILIC)
  normal phase

Description

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

apHera is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?