• USA Home
  • 56661-U - Micro-Mixer Static Mixers (2 - 25 μL)

EMAIL THIS PAGE TO A FRIEND
56661-U Supelco

Micro-Mixer Static Mixers (2 - 25 μL)

2 μL cartridge, stainless steel

  •  NACRES SB.51

Purchase

Properties

Related Categories ASI Static Mixers, Analytical/Chromatography, HPLC Accessories, HPLC/UHPLC, Micro-Mixer Static Mixers (2-25 μL) More...
material   stainless steel
compatibility   for use with ASI Micro Mixer Housing

Description

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?