• USA Home
  • 567238-U - Discovery® BIO Wide Pore C5 HPLC Column

567238-U Supelco

Discovery® BIO Wide Pore C5 HPLC Column

10 μm particle size, L × I.D. 25 cm × 21.2 mm

  •  eCl@ss 32110501



Related Categories 10 micron Discovery BIO Wide Pore (300) C5 Columns, Analytical/Chromatography, Discovery BIO, Discovery BIO HPLC Columns, Discovery BIO Wide Pore (300 Å) C5,
feature   endcapped
extent of labeling   3.5% Carbon loading
application(s)   HPLC: suitable
L × I.D.   25 cm × 21.2 mm
surface coverage   4.5 μmol/m2
matrix   silica particle platform
matrix active group   C5 (pentyl) phase
particle size   10 μm
pore size   300 Å
surface area   100 m2/g
mode of chromatography   reversed phase


General description

Discovery BIO Wide Pore C5 was designed for the efficient and reliable separation of proteins and peptides, especially hydrophobic peptides, by RP- HPLC. Long-chain phases, like C8 or C18, are often too hydrophobic for proteins and can cause excessively long retention time or even irreversible binding to the column. For this reason short-chain phases, typically C3 or C4, are often used for RP-HPLC of proteins. However, these short-chain phases are susceptible to hydrolysis, resulting in short column lifetime, especially at low pH. The Discovery BIO Wide Pore C5 gives elution order similar to a conventional C4, yet has enhanced pH stability for longer column lifetime. Generally, higher efficiency separations are achievable on the Discovery BIO Wide Pore C5 because of the improvements we have made to the silica and bonded-phase chemistry.

Legal Information

Discovery is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Milli-Q® Water Purification Solutions
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.


Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?