• USA Home
  • 567417-U - Discovery® HS PEG HPLC Column

567417-U Supelco

Discovery® HS PEG HPLC Column

5 μm particle size, L × I.D. 25 cm × 4.6 mm

  •  eCl@ss 32110501



Related Categories Analytical/Chromatography, Discovery HPLC Columns, Discovery HS PEG, HPLC, HPLC Columns, by Brand for Small Molecules,
feature   endcapped: no
extent of labeling   12% Carbon loading
parameter   ≤70 °C temp. range
L × I.D.   25 cm × 4.6 mm
surface coverage   3.8 μmol/m2
impurities   <10 ppm metals
matrix   silica gel, high purity, spherical particle platform
matrix active group   polyethyleneglycol phase
particle size   5 μm
pore size   120 Å
surface area   300 m2/g
operating pH range   2 - 8
mode of chromatography   reversed phase


General description

Discovery® HS PEG bonded phase provides reversed-phase separations that are distinctly different from C18 columns. It is an ideal candidate to choose when C18 columns give too much retention, when there is too much wasted space between peaks, or when you want to convert a gradient to an isocratic separation.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Milli-Q® Water Purification Solutions
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.


Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

guard cartridge

Product #


Add to Cart

567477-U Discovery® HS PEG Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 1 kit

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?