• USA Home
  • 567578-U - Discovery® HS F5 Supelguard Cartridge

567578-U Supelco

Discovery® HS F5 Supelguard Cartridge

5 μm particle size, L × I.D. 1 cm × 10 mm

  •  eCl@ss 32110501



Related Categories Analytical/Chromatography, Discovery HPLC Columns, Discovery HS F5, Discovery HS F5 Prep Columns, HPLC,
description   Supelguard Cartridge
packaging   pkg of 1 ea
application(s)   HPLC: suitable
L × I.D.   1 cm × 10 mm
matrix active group   PFP (pentafluorophenyl) phase
particle size   5 μm
pore size   120 Å
mode of chromatography   hydrophilic interaction (HILIC)
  reversed phase


Legal Information

Discovery is a registered trademark of Sigma-Aldrich Co. LLC

Supelguard is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Milli-Q® Water Purification Solutions
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.


Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?