• USA Home
  • 568270-U - Discovery® BIO Wide Pore C18 Supelguard Cartridge
568270-U Supelco
Discovery® BIO Wide Pore C18 Supelguard Cartridge
5 μm particle size, L × I.D. 2 cm × 2.1 mm
  •  eCl@ss 32110501
General description
Kits include one cartridge, a stand-alone holder, a piece of tubing, and 2 nuts and ferrules. Guard cartridges require holders that are sold separately. The 2.1 and 4 mm I.D. cartridges use 21150AST or 59660-U (both stand-alone) or 504254 or 55205 (both integral). The 10 mm I.D. cartridges use 567499-U. The 21.2 mm I.D. cartridges use 581392-U.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Discovery is a registered trademark of Sigma-Aldrich Co. LLC
Supelguard is a trademark of Sigma-Aldrich Co. LLC
Price and Availability
Milli-Q® Water Purification Solutions
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Certificate of Analysis
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?