• USA Home
  • 568312-U - Discovery® BIO Wide Pore C8 HPLC Column

568312-U Supelco

Discovery® BIO Wide Pore C8 HPLC Column

5 μm particle size, L × I.D. 15 cm × 4 mm

  •  eCl@ss 32110501



Related Categories 5 micron Discovery BIO Wide Pore (300) C8 Columns, Analytical/Chromatography, C8 HPLC Columns, Discovery BIO, Discovery BIO Wide Pore (300 Å) C8,
feature   endcapped
extent of labeling   5% Carbon loading
application(s)   HPLC: suitable
L × I.D.   15 cm × 4 mm
surface coverage   4 μmol/m2
matrix   silica particle platform
matrix active group   C8 (octyl) phase
particle size   5 μm
pore size   300 Å
surface area   100 m2/g
suitability   suitable for L7 per USP
mode of chromatography   reversed phase


General description

Discovery BIO Wide Pore C8 exhibits hydrophobicity intermediate between the Discovery BIO Wide Pore C5 and the Discovery BIO Wide Pore C18. The difference in hydrophobicity gives it unique selectivity relative to these other phases. It is ideal for peptide mapping because it provides complementary information compared to a C18 separation. Because of its intermediate hydrophobicity, we also recommend it for method development or scouting work. As with all Discovery BIO Wide Pore phases, the C8 phase gives efficient, symmetrical peaks, exceptional stability, long column lifetime, and LC-MS compatibility.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Milli-Q® Water Purification Solutions
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.


Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

guard cartridge

Product #


Add to Cart

568373-U Discovery® BIO Wide Pore C8 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 1 kit

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?