• USA Home
  • 568540-U - Discovery® HS C18 HPLC Column

568540-U Supelco

Discovery® HS C18 HPLC Column

5 μm particle size, L × I.D. 5 cm × 21.2 mm

  •  eCl@ss 32110501



Related Categories Analytical/Chromatography, C18 Columns, 5 micron, Discovery HPLC Columns, Discovery HS C18, Discovery HS C18 Prep Column, 5 micron,
feature   endcapped
extent of labeling   20% Carbon loading
parameter   ≤70 °C temp. range
application(s)   HPLC: suitable
L × I.D.   5 cm × 21.2 mm
surface coverage   3.2 μmol/m2
impurities   <10 ppm metals
matrix   silica gel, high purity, spherical particle platform
matrix active group   C18 (octadecyl) phase
particle size   5 μm
pore size   120 Å
surface area   300 m2/g
operating pH range   2 - 8
suitability   suitable for L1 per USP
mode of chromatography   reversed phase


Features and Benefits

• Stable, low bleed for LC-MS applications
• Scalable from analytical to preparatory
• Highly stable to ensure excellent run-to-run and lot-to-lot reproducibility
• Higher hydrophobicity for better resolution of difficult analytes

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Milli-Q® Water Purification Solutions
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.


Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?