• USA Home
  • 568574-U - Discovery® HS C18 Supelguard Guard Cartridge

568574-U Supelco

Discovery® HS C18 Supelguard Guard Cartridge

5 μm particle size, L × I.D. 1 cm × 10 mm, pkg of 1 ea

  •  eCl@ss 32110501



Related Categories Analytical/Chromatography, Discovery HPLC Columns, Discovery HS C18, Discovery HS C18 Prep Columns, Discovery HS C18 Supelguard Guard Cartridge,
packaging   pkg of 1 ea
application(s)   HPLC: suitable
L × I.D.   1 cm × 10 mm
matrix active group   C18 (octadecyl) phase
particle size   5 μm
pore size   120 Å
suitability   suitable for L1 per USP
mode of chromatography   reversed phase


Legal Information

Discovery is a registered trademark of Sigma-Aldrich Co. LLC

Supelguard is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Milli-Q® Water Purification Solutions
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.


Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?